Learn More
BioVendor, LLC Albumin Urinary Rat (S-type) Immunoturbidimetric Assay for autoanalyzers (60 tests)
SDP
Catalog No. 50893712
Encompass
Albumin Urinary Rat (S-type) Assay (60 tests). Immunoturbidimetric research reagent for an autoanalyzer to measure albumin in rat urine or serum using anti-rat albumin antibody. SAMPLE REQ: 6 l/well. CALIBRATION RANGE: 6.17500 g/ml. SUMMARY: Routine measurement of serum albumin is conveniently made with Shibayagis TIA assay kit for automatic analyzers. This assay can measure urinary albumin with high sensitivity, and is also applied to in vitro albumin biosynthesis system, checking of contamination with albumin in biological active substance preparations obtained by culture system, and liver transplantation experiments using NAR. Albumin is mostly a simple hydrophilic protein present in cells and body fluids. Albumin is synthesized in the liver, and serum albumin occupies 56-60% of total serum proteins. In normal human subjects, excretion of albumin into urine is low, about 30mg/day, but is increased in renal damage/disease. Manufactured by FUJIFILM Wako Shibayagi Corp., Japan.
Catalog No. 50-893-712 Supplier BioVendor, LLC Supplier No. 29392501
Albumin Urinary Rat (S-type) Assay (60 tests). Immunoturbidimetric research reagent for an autoanalyzer to measure albumin in rat urine or serum using anti-rat albumin antibody. SAMPLE REQ: 6 l/well. CALIBRATION RANGE: 6.17500 g/ml. SUMMARY: Routine measurement of serum albumin is conveniently made with Shibayagis TIA assay kit for automatic analyzers. This assay can measure urinary albumin with high sensitivity, and is also applied to in vitro albumin biosynthesis system, checking of contamination with albumin in biological active substance preparations obtained by culture system, and liver transplantation experiments using NAR. Albumin is mostly a simple hydrophilic protein present in cells and body fluids. Albumin is synthesized in the liver, and serum albumin occupies 56-60% of total serum proteins. In normal human subjects, excretion of albumin into urine is low, about 30mg/day, but is increased in renal damage/disease. Manufactured by FUJIFILM Wako Shibayagi Corp., Japan.