Learn More
Rockland Immunochemicals ELISA Microwell Blocking Buffer with Stabilizer (Azide and Mercury Free) 1 L
SDP
Supplier:  Rockland Immunochemicals MB0641000
Encompass
ELISA Microwell Blocking Buffer with Stabilizer is designed to block ELISA microwells coated with antigens antibodies or other ligands and to stabilize the plates for drying.  ELISA stabilizer allows the user to dry and store the plates for a minimum of one (1) year without significant loss of signal.
Catalog No. 50-194-3977