Learn More
BioVendor, LLC Elisa Chemerin Human BioVendor LLC
SDP
Supplier:  BioVendor, LLC RD191136200R
Encompass
TypeSandwich ELISA, Biotin-labelled antibodyApplicationsSerum, PlasmaSample Requirements5 ?l/wellShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore the complete kit at 2¿8¿C. Under these conditions, the kit is stable until the expiration date (see label on the box). Calibration Range0.25¿8 ng/mlLimit of Detection0.1 ng/ml
Catalog No. 50-894-205