Learn More
TRIBIOSCIENCE INC ELISA Coating Carbonate-Bicarbonate Buffer (pH9.6), 500 ML
SDP
Catalog No. NC3396060
Encompass
The ELISA Coating Carbonate-Bicarbonate Buffer (pH 9.6), 500 mL, is a high-quality buffer solution designed for optimal antigen or antibody coating in ELISA applications.
Catalog No. NC3396060 Supplier TRIBIOSCIENCE INC Supplier No. TBS5058
The ELISA Coating Carbonate-Bicarbonate Buffer (pH 9.6), 500 mL, is a high-quality buffer solution designed for optimal antigen or antibody coating in ELISA applications.