Learn More
BioVendor, LLC Elisa Fgf-21 Mouse/Rat BioVendor LLC
SDP
Supplier:  BioVendor, LLC RD291108200R
Encompass
TypeSandwich ELISA, Biotin-labelled antibodyApplicationsSerumSample Requirementsmouse samples: 7 ¿l/wellrat samples: 50 ¿l/wellShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore the complete kit at 2¿8¿C. Under these conditions, the kit is stable until the expiration date (see label on the box). Calibration Range40¿2560 pg/mlLimit of Detection18.4 pg/ml
Catalog No. NC9903102