Learn More
BioVendor, LLC Elisa Human Ngal BioVendor LLC
SDP
Supplier:  BioVendor, LLC RD191102200R
Encompass
TypeSandwich ELISA, Biotin-labelled antibodyApplicationsStool, Serum, Plasma-EDTA, UrineSample RequirementsSerum and plasma: 10 ¿l/wellFor processing the stool samples we recommend using BioVendor Extraction Buffer. The reagent is not included and can be ordered separately (Cat. No.: C005821, 100 ml).ShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore the kit at 2¿8¿C. Under these conditions, the kit is stable until the expiration date (see label on the box). Calibration Range0.3¿10 ng/mlLimit of Detection0.02 ng/ml
Catalog No. 50-871-223