Learn More
BioVendor, LLC ELISA HUMAN OPG 96W
SDP
Supplier:  BioVendor, LLC RD194003200
Encompass
Osteoprotegerin, TypeSandwich ELISA, Biotin-labelled antibodyApplicationsSerum, Plasma-EDTA, Plasma-Heparin, Plasma-CitrateSample Requirements35 µl/wellShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below.Storage/ExpirationStore the kit at 2–8°C. Under these conditions, the kit is stable until the expiration date (see label on the box). Calibration Range1.5–60 pmol/lLimit of Detection0.03 pmol/l
Catalog No. 50-871-226