Learn More
BioVendor, LLC Immunoglobulin E (total IgE) Mouse ELISA (96 wells)
SDP
Supplier:  BioVendor, LLC 29591101
Encompass
TypeSandwich ELISA, Biotin-labelled antibodyApplicationsSerum, PlasmaSample Requirements5 µl/wellShippingAt ambient temperature. Upon receipt, store the product at the temperature recommended below. Calibration Range1–100 ng/mlLimit of Detection50 pg/ml SUMMARY: IgE is a glycoprotein with a biological half life of about 3 days, and its blood level in normal human subject is low, about 300 ng/ml. B
Catalog No. 50-893-011