Learn More
Biomatik Corporation Phospho-Ret (Tyr1062) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51558
Encompass_Preferred
Phospho-Ret (Tyr1062) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Ret (Tyr1062) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P07949. Gene ID: 5979. Target Synonyms: C-ret; kinase Ret; Proto-oncogene ret precursor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9077