Learn More
Biomatik Corporation Phospho-Amyloid β A4 (Thr743/668) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50968
Encompass_Preferred
Phospho-Amyloid β A4 (Thr743/668) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Amyloid β A4 (Thr743/668) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P05067. Gene ID: 351. Target Synonyms: Amyloid beta A4 protein; ABPP; APPI; APP; Alzheimer disease amyloid protein; Cerebral vascular amyloid peptide; CVAP; PreA4; Protease nexin-II; PN-II; APP; A4; AD1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1184