Learn More
Biomatik Corporation Phospho-Keratin 8 (Ser73) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50979
Encompass_Preferred
Phospho-Keratin 8 (Ser73) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Keratin 8 (Ser73) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P05787. Gene ID: 3856. Target Synonyms: CK 8; CK8; CYK8; Cytokeratin endo A; K2C8; K8; KRT2-8; KRT8; Keratin type II cytoskeletal 8; cytokeratin 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1195