Learn More
Biomatik Corporation Phospho-Cyclin D3 (Thr283) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51059
Encompass_Preferred
Phospho-Cyclin D3 (Thr283) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Cyclin D3 (Thr283) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P30281. Gene ID: 896. Target Synonyms: CCND3; CGD3; Cyl-3; G1/S-specific cyclin D3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1260