Learn More
Biomatik Corporation Phospho-IR (Tyr1361) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291306
Encompass_Preferred
Phospho-IR (Tyr1361) Colorimetric Cell-Based ELISA Kit for detecting Phospho-IR (Tyr1361) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P06213. Gene ID: 3643. Target Synonyms: CD220 antigen; IR; insulin receptor; kinase InsR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1306 Supplier Biomatik Corporation Supplier No. EKA51111
Phospho-IR (Tyr1361) Colorimetric Cell-Based ELISA Kit for detecting Phospho-IR (Tyr1361) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P06213. Gene ID: 3643. Target Synonyms: CD220 antigen; IR; insulin receptor; kinase InsR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.