Learn More
Biomatik Corporation Phospho-MYB (Ser12) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291319
Encompass_Preferred
Phospho-MYB (Ser12) Colorimetric Cell-Based ELISA Kit for detecting Phospho-MYB (Ser12) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P10242. Gene ID: 4602. Target Synonyms: C-myb; Myb proto-oncogene protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1319 Supplier Biomatik Corporation Supplier No. EKA51125
Phospho-MYB (Ser12) Colorimetric Cell-Based ELISA Kit for detecting Phospho-MYB (Ser12) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P10242. Gene ID: 4602. Target Synonyms: C-myb; Myb proto-oncogene protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.