Learn More
Biomatik Corporation Phospho-PPAR-γ (Ser112) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291334
Encompass_Preferred
Phospho-PPAR-γ (Ser112) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PPAR-γ (Ser112) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P37231. Gene ID: 5468. Target Synonyms: NR1C3; PPAR-gamma-1; Peroxisome proliferator activated receptor gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1334 Supplier Biomatik Corporation Supplier No. EKA51147
Phospho-PPAR-γ (Ser112) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PPAR-γ (Ser112) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P37231. Gene ID: 5468. Target Synonyms: NR1C3; PPAR-gamma-1; Peroxisome proliferator activated receptor gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.