Learn More
Biomatik Corporation Phospho-ALOX5 (Ser271) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291400
Encompass_Preferred
Phospho-ALOX5 (Ser271) Colorimetric Cell-Based ELISA Kit for detecting Phospho-ALOX5 (Ser271) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P09917. Gene ID: 240. Target Synonyms: 5-lipoxygenase; ALOX5; Arachidonate 5-lipoxygenase; LOG5; LOX5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1400 Supplier Biomatik Corporation Supplier No. EKA51226
Phospho-ALOX5 (Ser271) Colorimetric Cell-Based ELISA Kit for detecting Phospho-ALOX5 (Ser271) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P09917. Gene ID: 240. Target Synonyms: 5-lipoxygenase; ALOX5; Arachidonate 5-lipoxygenase; LOG5; LOX5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.