Learn More
Biomatik Corporation Phospho-DAPK3 (Thr265) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51288
Encompass_Preferred
Phospho-DAPK3 (Thr265) Colorimetric Cell-Based ELISA Kit for detecting Phospho-DAPK3 (Thr265) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O43293. Gene ID: 1613. Target Synonyms: DAP kinase 3; DAP- like kinase; Death-associated protein kinase 3; Dlk; ZIP-kinase; ZIPK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1456