Learn More
Biomatik Corporation Phospho-ERK3 (Ser189) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51297
Encompass_Preferred
Phospho-ERK3 (Ser189) Colorimetric Cell-Based ELISA Kit for detecting Phospho-ERK3 (Ser189) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q16659. Gene ID: 5597. Target Synonyms: ERK-3; ERK3; Extracellular signal- regulated kinase 3; MAP kinase isoform p97; MAPK6; MK06; Mitogen-activated protein kinase 6; PRKM4; PRKM6; kinase ERK3; p55-MAPK; p97-MAPK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1464