Learn More
Biomatik Corporation Phospho-GluR4 (Ser862) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291468
Encompass_Preferred
Phospho-GluR4 (Ser862) Colorimetric Cell-Based ELISA Kit for detecting Phospho-GluR4 (Ser862) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P48058. Gene ID: 2893. Target Synonyms: GLR4; GLUR4; GRIA4; GluR-4; GluR-D; GluR4; Glutamate receptor 4 precursor; Glutamate receptor ionotropic; AMPA 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1468 Supplier Biomatik Corporation Supplier No. EKA51301
Phospho-GluR4 (Ser862) Colorimetric Cell-Based ELISA Kit for detecting Phospho-GluR4 (Ser862) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P48058. Gene ID: 2893. Target Synonyms: GLR4; GLUR4; GRIA4; GluR-4; GluR-D; GluR4; Glutamate receptor 4 precursor; Glutamate receptor ionotropic; AMPA 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.