Learn More
Biomatik Corporation Phospho-CDC16/APC6 (Ser560) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291491
Encompass_Preferred
Phospho-CDC16/APC6 (Ser560) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CDC16/APC6 (Ser560) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13042. Gene ID: 8881. Target Synonyms: APC6; Anaphase promoting complex subunit 6; CDC16Hs; Cell division cycle protein 16 homolog; Cyclosome subunit 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1491 Supplier Biomatik Corporation Supplier No. EKA51327
Phospho-CDC16/APC6 (Ser560) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CDC16/APC6 (Ser560) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13042. Gene ID: 8881. Target Synonyms: APC6; Anaphase promoting complex subunit 6; CDC16Hs; Cell division cycle protein 16 homolog; Cyclosome subunit 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.