Learn More
Biomatik Corporation Phospho-IL-2Rβ (Tyr364) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51340
Encompass_Preferred
Phospho-IL-2Rβ (Tyr364) Colorimetric Cell-Based ELISA Kit for detecting Phospho-IL-2Rβ (Tyr364) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P14784. Gene ID: 3560. Target Synonyms: CD122 antigen; High affinity IL-2 receptor beta subunit; IL-2 receptor; IL-2R beta; Interleukin-2 receptor beta chain precursor; P70-75. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1504