Learn More
Biomatik Corporation Phospho-ATP1A1 (Ser23) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51357
Encompass_Preferred
Phospho-ATP1A1 (Ser23) Colorimetric Cell-Based ELISA Kit for detecting Phospho-ATP1A1 (Ser23) in researchers plate their cell line of choice. Species Reactivity: Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P05023. Gene ID: 24211. Target Synonyms: A1A1; AT1A1; ATPase; Na+/K+ transporting; alpha 1 polypeptide; Hypothetical protein MGC38419; Na+/K+ ATPase 1; Na,K-ATPase 1; Sodium pump 1; Sodium/potassium-transporting ATPase alpha-1 chain precursor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1520