Learn More
Biomatik Corporation Phospho-PDCD4 (Ser457) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51369
Encompass_Preferred
Phospho-PDCD4 (Ser457) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PDCD4 (Ser457) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q53EL6. Gene ID: 27250. Target Synonyms: Programmed cell death protein 4; Nuclear antigen H731-like; Neoplastic transformation inhibitor protein; Protein 197/15a; PDCD4; H731. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1529