Learn More
Biomatik Corporation Phospho-Casein Kinase II α (Tyr255) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51374
Encompass_Preferred
Phospho-Casein Kinase II α (Tyr255) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Casein Kinase II α (Tyr255) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P68400. Gene ID: 1457. Target Synonyms: CK II; CK2-alpha1; CK2A1; CKIIA; CSK21; CSNK2A1; Casein kinase II; alpha chain; kinase CK2-alpha1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1534