Learn More
Biomatik Corporation Phospho-ARHGEF2 (Ser885) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51381
Encompass_Preferred
Phospho-ARHGEF2 (Ser885) Colorimetric Cell-Based ELISA Kit for detecting Phospho-ARHGEF2 (Ser885) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q92974. Gene ID: 9181. Target Synonyms: GEF-H1; GEFH1; Guanine nucleotide exchange factor GEF-H1; proliferating cell nucleolar antigen p40; rho/rac guanine nucleotide exchange factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1539