Learn More
Biomatik Corporation Phospho-p44/42 MAP Kinase (Thr202) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51442
Encompass_Preferred
Phospho-p44/42 MAP Kinase (Thr202) Colorimetric Cell-Based ELISA Kit for detecting Phospho-p44/42 MAP Kinase (Thr202) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P27361; P28482. Gene ID: 5595; 5594. Target Synonyms: ERK-1; ERK1; ERT2; Extracellular signal- regulated kinase 1; Insulin-stimulated MAP2 kinase; kinase ERK1; MAP kinase 1; MAPK 1; MAPK3; Microtubule- associated protein-2 kinase; Mitogen-activated protein kinase 3; MK03; MNK1; p44-ERK1; P44-ERK1; p44-MAPK; P44-MAPK; PRKM3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1586