Learn More
Biomatik Corporation Phospho-FOXO4 (Ser197) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291597
Encompass_Preferred
Phospho-FOXO4 (Ser197) Colorimetric Cell-Based ELISA Kit for detecting Phospho-FOXO4 (Ser197) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P98177. Gene ID: 4303. Target Synonyms: AFX; AFX1; Afxh; forkhead box protein O4; forkhead domain AFX1; MLLT7; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog; Drosophila) translocated to; 7 (AFX1). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1597 Supplier Biomatik Corporation Supplier No. EKA51456
Phospho-FOXO4 (Ser197) Colorimetric Cell-Based ELISA Kit for detecting Phospho-FOXO4 (Ser197) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P98177. Gene ID: 4303. Target Synonyms: AFX; AFX1; Afxh; forkhead box protein O4; forkhead domain AFX1; MLLT7; myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog; Drosophila) translocated to; 7 (AFX1). Restrictions: For Research Use Only. Not for use in diagnostic procedures.