Learn More
Biomatik Corporation Phospho-GluR2 (Ser880) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51462
Encompass_Preferred
Phospho-GluR2 (Ser880) Colorimetric Cell-Based ELISA Kit for detecting Phospho-GluR2 (Ser880) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P42262. Gene ID: 2891. Target Synonyms: Glutamate receptor 2; GluR-2; GluR-B; GluR-K2; Glutamate receptor ionotropic AMPA 2; AMPA-selective glutamate receptor 2; GLUR2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1603