Learn More
Biomatik Corporation Phospho-HNF4α (Ser313) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291613
Encompass_Preferred
Phospho-HNF4α (Ser313) Colorimetric Cell-Based ELISA Kit for detecting Phospho-HNF4α (Ser313) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P41235. Gene ID: 3172. Target Synonyms: HNF-4; HNF-4-alpha; HNF4; HNF4A; Hepatocyte nuclear factor 4-alpha; NR2A1; TCF14; Transcription factor 14; Transcription factor HNF-4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1613 Supplier Biomatik Corporation Supplier No. EKA51474
Phospho-HNF4α (Ser313) Colorimetric Cell-Based ELISA Kit for detecting Phospho-HNF4α (Ser313) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P41235. Gene ID: 3172. Target Synonyms: HNF-4; HNF-4-alpha; HNF4; HNF4A; Hepatocyte nuclear factor 4-alpha; NR2A1; TCF14; Transcription factor 14; Transcription factor HNF-4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.