Learn More
Biomatik Corporation Phospho-NF-κB p100/p52 (Ser865) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51517
Encompass_Preferred
Phospho-NF-κB p100/p52 (Ser865) Colorimetric Cell-Based ELISA Kit for detecting Phospho-NF-κB p100/p52 (Ser865) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q00653. Gene ID: 4791. Target Synonyms: DNA-binding factor KBF2; H2TF1; Lymphocyte translocation chromosome 10; Lyt10; NFKB2; Nuclear factor NF-kappa-B p100 subunit; Oncogene Lyt-10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1652