Learn More
Biomatik Corporation Phospho-NF-κB p105/p50 (Ser893) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51519
Encompass_Preferred
Phospho-NF-κB p105/p50 (Ser893) Colorimetric Cell-Based ELISA Kit for detecting Phospho-NF-κB p105/p50 (Ser893) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P19838. Gene ID: 4790. Target Synonyms: DNA-binding factor KBF1; EBP-1; NF-kappa-B1 p84/NF-kappa-B1 p98; NFKB1; NFkB-p50; Nuclear factor NF-kappa-B p105 subunit; nuclear factor NF-kappa-B p50 subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1654