Learn More
Biomatik Corporation Phospho-NMDAR1 (Ser897) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291660
Encompass_Preferred
Phospho-NMDAR1 (Ser897) Colorimetric Cell-Based ELISA Kit for detecting Phospho-NMDAR1 (Ser897) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q05586. Gene ID: 2902. Target Synonyms: GLURZ1; GRIN1; Glutamate [NMDA] receptor subunit zeta 1 precursor; N-methyl-D-aspartate receptor; NMD-R1; NMDZ1; NMZ1; NR1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1660 Supplier Biomatik Corporation Supplier No. EKA51526
Phospho-NMDAR1 (Ser897) Colorimetric Cell-Based ELISA Kit for detecting Phospho-NMDAR1 (Ser897) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q05586. Gene ID: 2902. Target Synonyms: GLURZ1; GRIN1; Glutamate [NMDA] receptor subunit zeta 1 precursor; N-methyl-D-aspartate receptor; NMD-R1; NMDZ1; NMZ1; NR1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.