Learn More
Biomatik Corporation Phospho-p53 (Ser315) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51533
Encompass_Preferred
Phospho-p53 (Ser315) Colorimetric Cell-Based ELISA Kit for detecting Phospho-p53 (Ser315) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P04637. Gene ID: 7157. Target Synonyms: Antigen NY-CO-13; Cellular tumor antigen p53; Phosphoprotein p53; TP53; tumor protein p53; Tumor suppressor p53. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1667