Learn More
Biomatik Corporation Phospho-p73 (Tyr99) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51542
Encompass_Preferred
Phospho-p73 (Tyr99) Colorimetric Cell-Based ELISA Kit for detecting Phospho-p73 (Tyr99) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O15350. Gene ID: 7161. Target Synonyms: p53-like transcription factor; p53-related protein; P73 alpha protein; p73-alpha; TP73; TRP73; Tumor protein p73. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1675