Learn More
Biomatik Corporation Phospho-IκB-ε (Ser22) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291193
Encompass_Preferred
Phospho-IκB-ε (Ser22) Colorimetric Cell-Based ELISA Kit for detecting Phospho-IκB-ε (Ser22) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O00221. Gene ID: 4794. Target Synonyms: I-kappa-B-epsilon; IKBE; IkappaBepsilon; NF-kappa-BIE; NF-kappaB inhibitor epsilon; NFKBIE. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1193 Supplier Biomatik Corporation Supplier No. EKA50977
Phospho-IκB-ε (Ser22) Colorimetric Cell-Based ELISA Kit for detecting Phospho-IκB-ε (Ser22) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O00221. Gene ID: 4794. Target Synonyms: I-kappa-B-epsilon; IKBE; IkappaBepsilon; NF-kappa-BIE; NF-kappaB inhibitor epsilon; NFKBIE. Restrictions: For Research Use Only. Not for use in diagnostic procedures.