Learn More
Biomatik Corporation Phospho-Smad1 (Ser465) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50990
Encompass_Preferred
Phospho-Smad1 (Ser465) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Smad1 (Ser465) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q15797. Gene ID: 4086. Target Synonyms: BSP-1; BSP1; Dwarfin-A; Dwf-A; JV4-1; MADH1; MADR1; Mad-related protein 1; Mad1; Mothers against DPP homolog 1; Mothers against decapentaplegic homolog 1; Mothers-against-DPP-related-1; SMAD 1; Smad 1; Transforming growth factor- beta signaling protein-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1203