Learn More
Biomatik Corporation Phospho-IGF1R (Tyr1161) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51000
Encompass_Preferred
Phospho-IGF1R (Tyr1161) Colorimetric Cell-Based ELISA Kit for detecting Phospho-IGF1R (Tyr1161) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P08069; P06213. Gene ID: 3480. Target Synonyms: Insulin-like growth factor 1 receptor; Insulin-like growth factor I receptor; IGF-I receptor; CD221 antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1212