Learn More
Biomatik Corporation Phospho-POLR2A (Ser1619) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51030
Encompass_Preferred
Phospho-POLR2A (Ser1619) Colorimetric Cell-Based ELISA Kit for detecting Phospho-POLR2A (Ser1619) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P24928. Gene ID: 5430. Target Synonyms: DNA-directed RNA polymerase II largest subunit; RNA pol II largest subunit; RNA polymerase II 220 kd subunit; RPB1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1235