Learn More
Biomatik Corporation Phospho-VASP (Ser157) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291245
Encompass_Preferred
Phospho-VASP (Ser157) Colorimetric Cell-Based ELISA Kit for detecting Phospho-VASP (Ser157) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P50552. Gene ID: 7408. Target Synonyms: Vasodilator-stimulated phosphoprotein; VASP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1245 Supplier Biomatik Corporation Supplier No. EKA51042
Phospho-VASP (Ser157) Colorimetric Cell-Based ELISA Kit for detecting Phospho-VASP (Ser157) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P50552. Gene ID: 7408. Target Synonyms: Vasodilator-stimulated phosphoprotein; VASP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.