Learn More
Biomatik Corporation Phospho-CSFR (Tyr723) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51196
Encompass_Preferred
Phospho-CSFR (Tyr723) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CSFR (Tyr723) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P07333. Gene ID: 1436. Target Synonyms: c-fms; CD115; CSF-1-R; CSF1R; CSFMR; FMS; Fms proto-oncogene; kinase CSFR; M-CSFR; Macrophage colony stimulating factor I receptor precursor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9029