Learn More
Biomatik Corporation Phospho-Caldesmon (Ser789) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291289
Encompass_Preferred
Phospho-Caldesmon (Ser789) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Caldesmon (Ser789) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q05682. Gene ID: 800. Target Synonyms: CAD; CALD1; CDM; L-caldesmon; Non-muscle caldesmon. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1289 Supplier Biomatik Corporation Supplier No. EKA51091
Phospho-Caldesmon (Ser789) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Caldesmon (Ser789) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q05682. Gene ID: 800. Target Synonyms: CAD; CALD1; CDM; L-caldesmon; Non-muscle caldesmon. Restrictions: For Research Use Only. Not for use in diagnostic procedures.