Learn More
Biomatik Corporation Phospho-FGFR1 (Tyr766) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51103
Encompass_Preferred
Phospho-FGFR1 (Tyr766) Colorimetric Cell-Based ELISA Kit for detecting Phospho-FGFR1 (Tyr766) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P11362. Gene ID: 2260. Target Synonyms: bFGF-R; c-fgr; FGFBR; FGFR-1; FGR1; fibroblast growth factor receptor 1; FLG; FLT2; Fms-like tyrosine kinase-2; kinase FGFR1; MFR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1299