Learn More
Biomatik Corporation Phospho-GRK2 (Ser29) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291302
Encompass_Preferred
Phospho-GRK2 (Ser29) Colorimetric Cell-Based ELISA Kit for detecting Phospho-GRK2 (Ser29) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P25098. Gene ID: 156. Target Synonyms: ADRBK1; ARBK1; BARK; BARK1; beta-adrenergic receptor kinase 1; beta-ARK-1; G protein receptor kinase 2; G- protein coupled receptor kinase 2; kinase GRK2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1302 Supplier Biomatik Corporation Supplier No. EKA51107
Phospho-GRK2 (Ser29) Colorimetric Cell-Based ELISA Kit for detecting Phospho-GRK2 (Ser29) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P25098. Gene ID: 156. Target Synonyms: ADRBK1; ARBK1; BARK; BARK1; beta-adrenergic receptor kinase 1; beta-ARK-1; G protein receptor kinase 2; G- protein coupled receptor kinase 2; kinase GRK2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.