Learn More
Biomatik Corporation Phospho-LAT (Tyr191) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51119
Encompass_Preferred
Phospho-LAT (Tyr191) Colorimetric Cell-Based ELISA Kit for detecting Phospho-LAT (Tyr191) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O43561. Gene ID: 27040. Target Synonyms: 36 kDa phospho-tyrosine adaptor protein; Linker for activation of T cells; p36-38; pp36. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1314