Learn More
Biomatik Corporation Phospho-PAK1/2/3 (Ser144/141/139) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51139
Encompass_Preferred
Phospho-PAK1/2/3 (Ser144/141/139) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PAK1/2/3 (Ser144/141/139) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13153; Q13177; O75914. Gene ID: 5058; 5062; 5063. Target Synonyms: Gamma-PAK; P21-activated kinase 2; PAK 2; PAK-2; PAK65; PAKI; S6/H4 kinase; kinase PAK2; p21-activated kinase 2; p21-activated protein kinase IBeta-PAK; CDC42/RAC effector kinase PAK-B; OPHN3; Oligophrenin-3; P65-PAK; PAK 3; PAK-3; PAKB; STK4; Serine/threonine-protein kinase PAK 3; kinase PAK3; p21-activated kinase 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1329