Learn More
Biomatik Corporation Phospho-BLNK (Tyr96) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291364
Encompass_Preferred
Phospho-BLNK (Tyr96) Colorimetric Cell-Based ELISA Kit for detecting Phospho-BLNK (Tyr96) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q8WV28. Gene ID: 29760. Target Synonyms: B-cell linker protein; LY57; SLP-65; SLP65. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1364 Supplier Biomatik Corporation Supplier No. EKA51182
Phospho-BLNK (Tyr96) Colorimetric Cell-Based ELISA Kit for detecting Phospho-BLNK (Tyr96) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q8WV28. Gene ID: 29760. Target Synonyms: B-cell linker protein; LY57; SLP-65; SLP65. Restrictions: For Research Use Only. Not for use in diagnostic procedures.