Learn More
Biomatik Corporation Phospho-CDC25C (Thr48) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51185
Encompass_Preferred
Phospho-CDC25C (Thr48) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CDC25C (Thr48) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P30307. Gene ID: 995. Target Synonyms: CDC25M1; dual specificity phosphatase Cdc25C; M-phase inducer phosphatase 3; MPI1B; MPIP3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1366