Learn More
Biomatik Corporation Phospho-LKB1 (Thr189) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51194
Encompass_Preferred
Phospho-LKB1 (Thr189) Colorimetric Cell-Based ELISA Kit for detecting Phospho-LKB1 (Thr189) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q15831. Gene ID: 6794. Target Synonyms: Serine/threonine-protein kinase LKB1; Renal carcinoma antigen NY-REN-19. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1374