Learn More
Biomatik Corporation Phospho-p63 (Ser455) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291384
Encompass_Preferred
Phospho-p63 (Ser455) Colorimetric Cell-Based ELISA Kit for detecting Phospho-p63 (Ser455) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9H3D4. Gene ID: 8626. Target Synonyms: EEC3; KET; LMS; p51; p73H; p73L; SHFM4; TA p63 alpha; TP63; tumor protein p63. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1384 Supplier Biomatik Corporation Supplier No. EKA51206
Phospho-p63 (Ser455) Colorimetric Cell-Based ELISA Kit for detecting Phospho-p63 (Ser455) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9H3D4. Gene ID: 8626. Target Synonyms: EEC3; KET; LMS; p51; p73H; p73L; SHFM4; TA p63 alpha; TP63; tumor protein p63. Restrictions: For Research Use Only. Not for use in diagnostic procedures.