Learn More
Biomatik Corporation Phospho-14-3-3 θ/τ (Ser232) Colorimetric Cell-Based ELISA Kit
Catalog No. 502291395
Encompass_Preferred
Phospho-14-3-3 θ/τ (Ser232) Colorimetric Cell-Based ELISA Kit for detecting Phospho-14-3-3 θ/τ (Ser232) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P27348. Gene ID: 5350. Target Synonyms: 14-3-3 protein T-cell; 14-3-3 protein tau; 1433T; 143T; HS1 protein; YWHAQ. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-229-1395 Supplier Biomatik Corporation Supplier No. EKA51220
Phospho-14-3-3 θ/τ (Ser232) Colorimetric Cell-Based ELISA Kit for detecting Phospho-14-3-3 θ/τ (Ser232) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P27348. Gene ID: 5350. Target Synonyms: 14-3-3 protein T-cell; 14-3-3 protein tau; 1433T; 143T; HS1 protein; YWHAQ. Restrictions: For Research Use Only. Not for use in diagnostic procedures.